Learn More
Medchemexpress LLC tert-Butyl 11-aminoundecanoate | 220851-29-6 | 99.02% | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY130715
Encompass_Preferred
tert-Butyl 11-aminoundecanoate is a PROTAC linker used in the synthesis of various PROTACs. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins by connecting two ligands: one for an E3 ubiquitin ligase and another for the target protein. This product is for research use only.
  • PROTAC linker
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-42671