Learn More
Medchemexpress LLC Tert-butyl 2-(2-(2-(2-(4-aminophenoxy)ethoxy)ethoxy)ethoxy)acetate | 2115897-27-1 | 355.43 g/mol | C18H29NO6 | 5 MG

Supplier: Medchemexpress LLC HY1646375MG
Aniline-PEG3-C1-Boc (compound D-1) is a Boc-protected aniline connected via a three-unit polyethylene glycol (PEG3) spacer and a one-carbon linker. It is used as a building-block intermediate for cytotoxic drug-linker polymers and for the synthesis of antibody-drug conjugates (ADCs).
- Used as an intermediate for cytotoxic drug-linker polymers and ADC synthesis.
- CAS number: 2115897-27-1.
- Molecular formula: C18H29NO6; molecular weight: 355.43 g/mol.
- Physical state: liquid.
- Typical pack size: 5 MG.
- Predicted density: 1.111 ± 0.06 g/cm³ (predicted).
- Refer to the supplier safety data sheet for storage, handling, and purity information.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.