Learn More
Medchemexpress LLC Tert-butyl 2-(2-(2-(2-hydroxyethoxy)ethoxy)ethoxy)ethyl carbamate | 106984-09-2 | MFCD07779195 | 99.5% | 293.36 g/mol | C13H27NO6 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW025896500MG
Encompass_Preferred
Boc-NH-PEG4 is a Boc-protected PEG4 linker used as a hydrophilic spacer in proteolysis-targeting chimera (PROTAC) synthesis and other bioconjugation chemistries. It combines a protected amine and a terminal hydroxyl to allow stepwise functionalization, improve aqueous solubility, and provide conformational flexibility during linker assembly.
  • Provides a Boc-protected amine for orthogonal deprotection and coupling.
  • Terminating hydroxyl enables diverse functionalization reactions.
  • PEG4 spacer increases solubility and molecular flexibility.
  • High purity supports reliable synthetic workflows (listed ~99.5%).
  • Molecular weight ~293.36 g/mol; formula C13H27NO6.
  • Supplied in laboratory-scale quantities for research use.
Catalog No. 50-003-01745