Learn More
Medchemexpress LLC Tert-butyl 2,2-dimethyl-5-oxopiperidine-1-carboxylate | 1894533-96-0 | MFCD31718688 | 97.2% | 227.3 g/mol | C12H21NO3 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW451756500MG
Encompass_Preferred
Tert-butyl 2,2-dimethyl-5-oxopiperidine-1-carboxylate is a Boc-protected piperidinone intermediate used in medicinal and synthetic chemistry for preparation of more complex heterocycles and active pharmaceutical candidates. It is supplied as a solid for research use and is intended for small-scale synthetic applications under standard laboratory handling conditions.
  • Used as a Boc-protected piperidinone intermediate for synthesis.
  • High purity suitable for research-scale synthetic work.
  • Molecular formula C12H21NO3 and molecular weight 227.3 g/mol.
  • Offered in small package sizes for laboratory use.
  • Stable at room temperature for extended storage when dry.
Catalog No. 50-002-81408