Learn More
Medchemexpress LLC Tert-butyl (2-(pyridin-2-yldisulfanyl)ethyl)carbamate | 535943-48-7 | MFCD24469377 | 96.0% | 286.42 | C12H18N2O2S2 | 250 MG
SDP
Supplier:  Medchemexpress LLC HYW578278250MG
Encompass_Preferred
tert-Butyl (2-(pyridin-2-yldisulfanyl)ethyl)carbamate is a Boc-protected carbamate containing a pyridinyl-disulfide functionality used as a thiol-reactive labeling reagent or intermediate in organic synthesis and protein modification studies. Intended for research use and supplied with analytical documentation.
  • Boc-protected carbamate functionality for amine protection.
  • Thiol-reactive pyridinyl-disulfide moiety for reversible cysteine labeling.
  • High purity suitable for analytical and preparative work.
  • Supplied with certificate of analysis and safety data sheet.
  • Recommended storage: 4°C under nitrogen; in solvent: -80°C (6 months), -20°C (1 month).
  • Available in multiple sizes including 250 mg.
Catalog No. 50-002-84148