Learn More
Medchemexpress LLC Tert-butyl (20-hydroxy-3,6,9,12,15,18-hexaoxaicosyl)carbamate | 1292268-13-3 | MFCD29049415 | 97.0% | 425.51 g/mol | C19H39NO9 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY130505100MG
Encompass_Preferred
N-Boc-PEG7-alcohol is a Boc-protected PEG7 linker terminating in a primary alcohol, used as a cleavable linker in antibody-drug conjugate (ADC) synthesis and as a spacer in PROTAC construction. The polyethylene glycol spacer provides hydrophilicity and flexibility for bioconjugation and medicinal chemistry applications.
  • Used for PROTAC and ADC linker synthesis.
  • Boc-protected amine with a PEG7 spacer terminating in a primary alcohol.
  • Available in multiple package sizes, including 100 MG.
  • High purity (≈97.0%) suitable for research use.
  • Follow recommended storage conditions for pure form and in solvent to preserve stability.
Catalog No. 50-002-03238