Learn More
Medchemexpress LLC Tert-butyl (26-hydroxy-3,6,9,12,15,18,21,24-octaoxahexacos-1-yl) carbamate | 2112731-44-7 | MFCD30723250 | >98.0% | 513.62 g/mol | C23H47NO11 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0715831G
Encompass_Preferred
N-Boc-PEG9-alcohol is a Boc-protected polyethylene glycol (PEG9) primary alcohol used as a cleavable linker in medicinal chemistry. It is commonly employed in the synthesis of proteolysis-targeting chimeras (PROTACs) and antibody-drug conjugates (ADCs) to provide a flexible, hydrophilic spacer between ligands and payloads.
  • Provides a Boc-protected primary alcohol functional group for downstream derivatization.
  • Contains a nine-unit PEG spacer to increase solubility and flexibility.
  • Suitable for use as a cleavable ADC or PROTAC linker in medicinal chemistry workflows.
  • Available in small-scale laboratory pack sizes for research synthesis.
Catalog No. 50-002-02931