Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tert-butyl 3-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]propionate | 883554-11-8 | MFCD26388566 | >98.0% | 292.37 g/mol | C14H28O6 | 500 MG
SDP

Supplier:  Medchemexpress LLC HY141395500MG

Encompass_Preferred

m-PEG4-Boc is a cleavable four-unit polyethylene glycol (PEG) linker with a tert-butyl (Boc) protected methyl terminus, designed for use in bioconjugation workflows such as antibody-drug conjugates (ADCs) and PROTACs.

  • Cleavable 4-unit PEG linker.
  • CAS number 883554-11-8.
  • Molecular formula C14H28O6.
  • Molecular weight 292.37 g/mol.
  • Purity >98.0%.
  • Typical package 500 mg.
  • Recommended storage: solid at -20°C for long-term storage.

Catalog No. 50-002-02223


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.