Learn More
Medchemexpress LLC Tert-butyl 3-(4-chloro-7H-pyrrolo[2,3-d]pyrimidin-7-yl)pyrrolidine-1-carboxylate | 1240301-66-9 | 96.2% | 322.80 | 100 MG

Supplier: Medchemexpress LLC HY22403100MG
tert-Butyl 3-(4-chloro-7H-pyrrolo[2,3-d]pyrimidin-7-yl)pyrrolidine-1-carboxylate is a drug intermediate for the synthesis of various active compounds. It is for research use only and not sold to patients.
- Solid appearance
- White to light yellow color
- Purity of 96.20%
- Available with data sheet, COA, SDS, handling instructions, HNMR, and LCMS documents
- Ships at room temperature in continental US
- Storage at -20°C in sealed conditions, away from moisture
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.