Learn More
Medchemexpress LLC tert-butyl (3-aminobicyclo[1.1.1]pentan-1-yl)carbamate | 1638767-25-5 | MFCD27987307 | 100.0% | 198.27 g/mol | C10H18N2O2 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW054424100G
Encompass_Preferred
tert-butyl (3-aminobicyclo[1.1.1]pentan-1-yl)carbamate is a Boc-protected primary amine of the bicyclo[1.1.1]pentane scaffold used as a synthetic intermediate in medicinal chemistry for construction of active compounds and linkers.
  • Boc-protected primary amine for downstream derivatization.
  • High reported purity, suitable for research and synthesis.
  • Compact, strained bicyclo[1.1.1]pentane core useful as a phenyl bioisostere.
  • Molecular formula C10H18N2O2; molecular weight ~198.27 g/mol.
  • Available in multiple pack sizes including 100 G.
Catalog No. 50-002-09950