Learn More
Medchemexpress LLC Tert-butyl 3-ethynylazetidine-1-carboxylate | 287193-01-5 | MFCD17016080 | 99.8% | 181.23 g/mol | C10H15NO2 | 1 G

Supplier: Medchemexpress LLC HYW0503671G
Tert-butyl 3-ethynylazetidine-1-carboxylate is an organic intermediate used in medicinal chemistry for the synthesis of nitrogen-containing heterocycles and other bioactive compounds.
- Used as a drug intermediate in organic synthesis.
- CAS number 287193-01-5.
- Molecular formula C10H15NO2; molecular weight 181.23 g/mol.
- Typical purity about 99.8%.
- Available in small pack sizes for research (250 mg-5 g); larger quantities available on request.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.