Learn More
Medchemexpress LLC Tert-butyl (3-iodopropyl)carbamate | 167479-01-8 | MFCD09951819 | >97.0% | 285.12 g/mol | C8H16INO2 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW053842500MG
Encompass_Preferred
tert-Butyl (3-iodopropyl)carbamate is a Boc-protected 3-iodopropyl carbamate used as an intermediate in organic synthesis. The terminal iodine serves as a good leaving group for nucleophilic substitution, and the Boc protecting group can be removed under mild acidic conditions to yield the free amine.
  • Used as a synthetic intermediate for introducing a 3-iodopropyl group.
  • Boc group removable under mild acidic conditions to provide the free amine.
  • Terminal iodine provides good leaving group reactivity for substitution reactions.
  • Offered with high purity suitable for research and synthesis applications.
  • Available in multiple small-scale sizes for lab use.
Catalog No. 50-002-10043