Learn More
Medchemexpress LLC Tert-butyl 3-iodopropylcarbamate | 167479-01-8 | ≥97.0% | 285.12 g/mol | C8H16INO2 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0538421G
Encompass_Preferred
tert-Butyl 3-iodopropylcarbamate is a Boc-protected 3-iodopropyl carbamate used as a synthetic building block and alkylating reagent. The terminal iodine serves as a good leaving group for nucleophilic substitution, and the tert-butoxycarbonyl (Boc) group can be removed under mild acidic conditions to reveal an amine.
  • Boc-protected 3-iodopropyl carbamate functional group
  • terminal iodine offers efficient leaving group for substitution
  • Boc group removable under mild acidic conditions
  • high purity suitable for synthetic applications
  • useful as an alkylating reagent and building block in medicinal chemistry
Catalog No. 50-002-10919