Learn More
Medchemexpress LLC Tert-butyl 3-iodopropylcarbamate | 167479-01-8 | MFCD09951819 | ≥97.0% | 285.12 | C8H16INO2 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYW053842
Encompass_Preferred
tert-Butyl 3-iodopropylcarbamate is an organic carbamate reagent used as an alkylating intermediate in organic synthesis and medicinal chemistry research. It is a solid compound with a molecular weight of 285.12 and typical purity ≥97.0% (NMR). Recommended storage is at -20 °C under inert atmosphere to preserve stability.
  • Used as a reactive alkylating building block in multi-step synthesis.
  • High analytical purity suitable for research applications.
  • Solid form allows easy weighing and handling for small-scale reactions.
  • Stable under recommended storage conditions (-20 °C, inert atmosphere).
  • Contains an iodopropyl leaving group useful for subsequent substitution chemistry.
Catalog No. 50-002-09605