Learn More
Medchemexpress LLC Tert-butyl 3-oxoazetidine-1-carboxylate | 398489-26-4 | 100.0% | 171.19 | 25 G

Supplier: Medchemexpress LLC 25GHY40146
tert-Butyl 3-oxoazetidine-1-carboxylate is a synthetic intermediate vital for the discovery and development of compounds targeting cancer and tuberculosis. This compound features a molecular weight of 171.19 and is available as a white to light yellow solid with high purity.
- Synthetic intermediate
- Used in the discovery and development of compounds targeting cancer and tuberculosis
- Molecular weight: 171.19
- CAS number: 398489-26-4
- Appearance: Solid
- Color: White to light yellow
- Purity: 99.99%
- Molecular formula: C8H13NO3
- Synonym: N-Boc-3-azetidinone
- Recommended storage: Powder at -20°C for 3 years, 4°C for 2 years; in solvent at -80°C for 6 months, -20°C for 1 month
- Solubility in DMSO: 100 mg/mL (584.15 mM)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.