Learn More
eMolecules tert-butyl (3R,4R)-3-amino-4-fluoropyrrolidine-1-carboxylate | 1441392-27-3 | | 1g
Supplier: eMolecules 551214316
Pharmablock | tert-butyl (3R,4R)-3-amino-4-fluoropyrrolidine-1-carboxylate | 1g | 551214316 | PB07377 | | 1441392-27-3 | | 204.245 | C9H17FN2O2
If you are unable to find the chemical you are looking for, make sure you are logged into your fishersci.com account and click on the following link:
eMolecules Building Block Tool