Learn More
Medchemexpress LLC tert-Butyl 4-formylpiperidine-1-carboxylate | 137076-22-3 | 99.5% | C11H19NO3 | 100 G

Supplier: Medchemexpress LLC HY69260100G
tert-Butyl 4-formylpiperidine-1-carboxylate is the linker of PROTAC BRM/BRG1 degrader-3 (HY-168251). It is for research use only.
- For research use only
- Molecular weight: 213.27
- Formula: C11H19NO3
- Appearance: White to yellow solid below 34°C, liquid above 40°C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.