Learn More
Medchemexpress LLC Tert-butyl 4-formylpiperidine-1-carboxylate | 137076-22-3 | 99.54% | C11H19NO3 | 50 G
SDP
Supplier:  Medchemexpress LLC HY6926050G
Encompass_Preferred
tert-Butyl 4-formylpiperidine-1-carboxylate is a chemical compound primarily known for its role as a linker. It is specifically identified as a component in a PROTAC BRM/BRG1 degrader, indicating its utility in targeted protein degradation research and development.
  • Functions as a linker molecule
  • Component in a PROTAC BRM/BRG1 degrader
  • Used in targeted protein degradation research
Catalog No. 50-004-34291