Learn More
Medchemexpress LLC Tert-butyl (4-iodobutyl)carbamate | 262278-40-0 | MFCD09951820 | 98.0% | 299.15 g/mol | C9H18INO2 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW092078500MG
Encompass_Preferred
Tert-butyl (4-iodobutyl)carbamate is a Boc-protected 4-iodobutyl carbamate used as a four-carbon linker in organic synthesis. The Boc protecting group can be removed under mild acidic conditions to reveal a free amine, and the iodine provides a reactive leaving group for nucleophilic substitution and further functionalization.
  • Four-carbon linker for introduction of Boc-protected aminoalkyl chains.
  • Boc group removable under mild acidic conditions to yield free amine.
  • Iodine atom enables nucleophilic substitution and cross-coupling reactions.
  • Commonly supplied at high purity for synthetic applications.
  • Available in multiple pack sizes to suit small-scale synthesis and scale-up.
Catalog No. 50-002-08715