Learn More
Medchemexpress LLC Tert-butyl 6-anilino pyrrolo[3,2-c]pyridine-1-carboxylate (chloro, imidazolyl, pyrazolyl substituted) | 1400284-80-1 | 99.1% | 503.98 g·mol⁻¹ | C26H26ClN7O2 | 50MG
SDP
Supplier:  Medchemexpress LLC HY1260350MG
Encompass_Preferred
CCT251455 is a potent and selective inhibitor of the mitotic kinase monopolar spindle 1 (MPS1), used in biochemical and cell-based research to probe mitotic checkpoint function and cell-cycle regulation. Supplied as a purified solid with accompanying documentation, it is intended for laboratory research use only and is available in multiple pack sizes and solution formats for convenience.
  • Potent, selective MPS1 kinase inhibitor (IC50 3 nM).
  • High purity suitable for research applications (99.1%).
  • Molecular weight 503.98 g·mol⁻¹ and formula C26H26ClN7O2.
  • Available as solid packs and as a 10 mM solution in DMSO.
  • Documentation provided: product data sheet, COA, SDS.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-36592