Learn More
Medchemexpress LLC tert-Butyl 6-bromohexanoate | 65868-63-5 | 99.7% | 251.16 | 25 G

Supplier: Medchemexpress LLC 25GHY43191
tert-Butyl 6-bromohexanoate is a PROTAC linker that can be used to design PROTACs, such as PROTAC ERK5 degrader-1. It is for research use only.
- Appearance: Liquid
- Color: Colorless to light yellow
- Shipping: Room temperature in continental US; may vary elsewhere
- Storage: Pure form: -20°C for 3 years, 4°C for 2 years; In solvent: -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.