Learn More
Medchemexpress LLC Tert-butyl (6-fluoro-4-(dioxaborolan-2-yl)-5-(triisopropylsilyl)ethynyl)naphthalen-2-yl carbamate | 2839207-57-5 | 99.6% | 567.61 | C32H47BFNO4Si | 100 G

Supplier: Medchemexpress LLC HYW599085100G
tert-Butyl (6-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5-((triisopropylsilyl)ethynyl)naphthalen-2-yl)carbamate is an organic intermediate used in medicinal chemistry for synthesis of active compounds. The material is supplied as a high-purity white solid with accompanying analytical and safety documentation.
- High purity (99.6%).
- Chemical formula C32H47BFNO4Si.
- Molecular weight 567.61 g/mol.
- White solid suitable as a drug intermediate.
- Available in 100 G packaging.
- Supplied with certificate of analysis and safety data sheet.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.