Learn More
Medchemexpress LLC Tert-butyl (6-fluoro-4-(pinacol boronate)-5-((triisopropylsilyl)ethynyl)naphthalen-2-yl)carbamate | 2839207-57-5 | MFCD22064227 | 99.8% | 567.61 g/mol | C32H47BFNO4Si | 25 G
SDP
Supplier:  Medchemexpress LLC HYW59908525G
Encompass_Preferred
tert-Butyl (6-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-5-((triisopropylsilyl)ethynyl)naphthalen-2-yl)carbamate is a fluoro-substituted tert-butyl carbamate bearing a pinacol boronate and a triisopropylsilyl-protected ethynyl; it is used as a drug intermediate in small-molecule synthesis.
  • High purity: 99.8% (HPLC) according to COA.
  • Functionalized building block containing a pinacol boronate for cross-coupling.
  • Triisopropylsilyl-protected ethynyl enables selective downstream reactions.
  • Stable when stored under inert atmosphere; see SDS for recommended conditions.
  • Supplied as a 25 G package suitable for batch synthesis.
Catalog No. 50-002-84462