Learn More
Medchemexpress LLC tert-Butyl (7-aminoheptyl)carbamate | 99733-18-3 | 97.0% | 230.35 | 5 G

Supplier: Medchemexpress LLC HY223915G
tert-Butyl (7-aminoheptyl)carbamate is a PROTAC linker used in the synthesis of CPD-10. This compound appears as a solid below 39°C and a liquid above 41°C, with a colorless to off-white appearance. It is typically shipped at room temperature in the continental US.
- Can be used to synthesize CPD-10.
- Appears as a solid below 39°C and a liquid above 41°C.
- Colorless to off-white appearance.
- Recommended storage at 4°C, protected from light.
- In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.