Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC tert-Butyl (7-aminoheptyl)carbamate | 99733-18-3 | ≥98.0% | 230.35 | 100 MG
SDP

Supplier:  Medchemexpress LLC HY22391100MG

Encompass_Preferred

tert-Butyl (7-aminoheptyl)carbamate is a PROTAC linker and can be used to synthesize CPD-10 (HY-168660).

  • PROTAC linker
  • Appearance: <39°C Solid, >41°C Liquid
  • Color: Colorless to off-white

Catalog No. 50-004-02099


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.