Learn More
Medchemexpress LLC tert-Butyl 9-aminon 250mg
SDP
Supplier:  Medchemexpress LLC 250MGHYW101722
Encompass_Preferred
tert-Butyl (9-aminononyl)carbamate is an alkane chain with terminal amine and Boc-protected amino groups The compound can be used as a PROTAC linker in the synthesis of PROTACs and other conjugation applications Amine group is reactive with carboxylic acids activated NHS esters carbonyls (ketone aldehyde) etc The Boc group can be deprotected under mild acidic conditions to form the free amine
Catalog No. 50-002-04655