Learn More
Medchemexpress LLC Tert-butyl hydrogen tetradecanedioate | 234082-00-9 | MFCD23136040 | 99.9% | 314.46 g/mol | C18H34O4 | 5 G

Supplier: Medchemexpress LLC HYW0749535G
Tert-butyl hydrogen tetradecanedioate is a small-molecule organic reagent (C18H34O4, 314.46 g/mol, CAS 234082-00-9) used as a biochemical assay reagent and synthetic intermediate for laboratory research. Store at room temperature for up to three years; in solvent, store at -80°C for two years or -20°C for one year.
- Small-molecule biochemical assay reagent and synthetic intermediate.
- Molecular weight 314.46 g/mol, formula C18H34O4.
- High purity (99.9% by HPLC) suitable for research use.
- Recommended storage: room temperature for 3 years; in solvent, -80°C for 2 years or -20°C for 1 year.
- Suitable for use in organic synthesis and assay development.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.