Learn More
Medchemexpress LLC Tert-butyl N-[3-(piperazin-1-yl)propyl]carbamate | 874831-60-4 | MFCD06808577 | >97.0% | 243.35 | C12H25N3O2 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0884565G
Encompass_Preferred
tert-Butyl N-[3-(piperazin-1-yl)propyl]carbamate is a small-molecule linker and synthetic intermediate commonly used in PROTAC construction and medicinal chemistry research. It is supplied at research-grade purity and offered in multiple sizes for lab-scale synthesis.
  • Used as a PROTAC linker and synthetic building block.
  • High purity suitable for research (typically ≥97%).
  • Liquid, colorless to light yellow appearance for easy handling.
  • Available in multiple lab-scale sizes, including 5 g.
  • Store protected from light at refrigerated or low-temperature conditions.
Catalog No. 50-002-08290