Learn More
Medchemexpress LLC tert-butyl N-(4-azidobutyl)carbamate | 129392-85-4 | MFCD24452574 | ≥98.0% | 214.26 | C9H18N4O2 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW4579535G
Encompass_Preferred
tert-Butyl N-(4-azidobutyl)carbamate is an N-Boc-protected azidobutyl carbamate used as a bifunctional crosslinker and synthetic building block for chemical biology and organic synthesis. The azide moiety enables bioorthogonal conjugation via azide-alkyne cycloaddition, and the Boc protecting group can be removed under mild acidic conditions to expose a free amine for further derivatization.
  • Enables copper-catalyzed or strain-promoted azide-alkyne cycloaddition for stable triazole formation.
  • Boc protection provides orthogonal protection and straightforward acid-mediated deprotection.
  • Suitable as a bifunctional crosslinker and building block for bioconjugation workflows.
  • Molecular formula C9H18N4O2; molecular weight 214.26 g/mol for reference.
Catalog No. 50-002-81500