Learn More
Medchemexpress LLC Tert-butyl N-(6-bromohexyl)carbamate | 142356-33-0 | MFCD06201019 | 98.0% | 280.20 g/mol | C11H22BrNO2 | 1 G

Supplier: Medchemexpress LLC HYW0115611G
Boc-NH-C6-Br (tert-butyl N-(6-bromohexyl)carbamate) is a non-cleavable linker used in the synthesis of antibody-drug conjugates (ADCs). It is provided for research use only and is intended for laboratory applications involving linker conjugation and small-molecule functionalization.
- Non-cleavable linker suitable for antibody-drug conjugates
- Molecular formula: C11H22BrNO2
- Molecular weight: 280.20 g/mol
- CAS number: 142356-33-0
- Purity: 98.0%
- Appearance: colorless to off-white solid-liquid mixture
- Available in multiple pack sizes, including 1 g and larger laboratory quantities
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.