Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC tert-Butyl (R)-2-((1S,2S)-3-(((1R,2S)-1-hydroxy-1-phenylpropan-2-yl)amino)-1-methoxy-2-methyl-3-oxopropyl)pyrrolidine-1-carboxylate | 2994330-72-0 | 97.1% | C23H36N2O5 | 50 MG
SDP

Supplier:  Medchemexpress LLC HY78931A50MG

Encompass_Preferred

MedChemExpress HY-78931A is a chemical compound with the molecular formula C23H36N2O5 and a molecular weight of 420.54. This white to off-white solid boasts a high purity of 97.13%. It is typically stored at 4°C under nitrogen, with specific recommendations for its storage in solvent.

  • White to off-white solid appearance
  • High purity of 97.13%
  • Structure consistent with 1H NMR spectrum
  • Complies with all given specifications
  • Recommended storage at 4°C under nitrogen
  • In-solvent storage: -80°C for 6 months, -20°C for 1 month (under nitrogen)

Catalog No. 50-004-35096


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.