Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tert-butyl ((S)-1-amino-3-((R)-tetrahydro-2H-pyran-3-yl)propan-2-yl)(methyl)carbamate | 942145-27-9 | 95.0% | 272.39 | 250 MG
SDP

Supplier:  Medchemexpress LLC 250MGHY32289

Encompass_Preferred

tert-Butyl ((S)-1-amino-3-((R)-tetrahydro-2H-pyran-3-yl)propan-2-yl)(methyl)carbamate is a drug intermediate used for the synthesis of various active compounds.

  • Appearance: liquid
  • Color: brown to black
  • Pure form storage: -20°C for 3 years; 4°C for 2 years
  • In solvent storage: -80°C for 6 months; -20°C for 1 month
  • For research purposes only

Catalog No. 50-004-02405


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.