Learn More
Medchemexpress LLC Tert-butyl (tert-butoxycarbonyl)-L-serinate | 7738-22-9 | MFCD00190832 | 98.0% | 261.32 g/mol | C12H23NO5 | 1 G

Supplier: Medchemexpress LLC HYW0418671G
Boc-Ser-OtBu is the N-tert-butoxycarbonyl protected tert-butyl ester of L-serine used as an amino acid derivative and synthetic intermediate in organic and peptide chemistry. It is supplied as a research reagent with a reported purity of 98.0%.
- Protected serine derivative suitable as a synthetic intermediate.
- High purity: 98.0%.
- Molecular formula C12H23NO5; molecular weight 261.32 g/mol.
- Soluble in DMSO (200 mg/mL); ultrasonic assistance recommended.
- Recommended storage: 4°C, protect from light; in solvent, -80°C (6 months) or -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.