Learn More
Medchemexpress LLC Tertiapin-Q | 910044-56-3 | 99.8% | 2452.00 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP12755MG
Encompass_Preferred
Tertiapin-Q is a highly selective blocker of GIRK1/4 heterodimer and ROMK1 (Kir1.1) potassium channels, intended for research use only. It also potently and selectively blocks Kir3.1-Kir3.4 channels and calcium-activated large conductance potassium channels, blocking somatostatin (SS-14)-activated current. As a muscarinic acetylcholine receptor-operated K+ current (IK,Ach) blocker, Tertiapin-Q significantly prolongs the atrial effective refractory period (AERP).
  • Highly selective blocker of GIRK1/4 heterodimer and ROMK1 (Kir1.1) potassium channels
  • Potently and selectively blocks Kir3.1-Kir3.4 and calcium-activated large conductance potassium channels
  • Blocks somatostatin (SS-14)-activated current
  • Functions as a muscarinic acetylcholine receptor-operated K+ current (IK,Ach) blocker
  • Prolongs atrial effective refractory period (AERP)
Catalog No. 50-004-23775