Learn More
Terumo BCT Inc TSCD-II Sterile Tubing Welder
Supplier:  Terumo BCT Inc 3MESC203A
Encompass
Designed for quality, reliability and flexibility, the TSCD-II welder is a next-generation, closed-system device that can be used to connect polyvinyl chloride (PVC) tubing in any combination of wet and dry.
Catalog No. NC0575379
Add to Cart
Request a Quote