Learn More
Medchemexpress LLC TES sodium | 70331-82-7 | 98.0% | 251.23 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHY23430A
Encompass_Preferred
TES sodium is an excellent hydrogen ion buffer with characteristics such as an appropriate dissociation constant, a low metal-binding constant, and high water solubility. It can be used in research involving mammalian cell culture, viruses, and similar applications.
  • Appropriate dissociation constant
  • Low metal-binding constant
  • High water solubility
  • Used in research involving mammalian cell culture
  • Used in research involving viruses
Catalog No. 50-004-02977