Learn More
MSC TESA Brown & Sharpe Magnetic Indicator Base, 1-1/4 in Base, Ball Socket Arm, 15 Lbs
Supplier:  MSC 06581771
Encompass
This magnetic indicator base provides stable support for precise measurements. It features a strong magnetic pull and a flexible arm, making it suitable for various laboratory and industrial applications. The base dimensions ensure a secure placement, and the included holders add convenience.
  • Magnetic pull of 15 Lbs
  • Post (ball & socket) arm style
  • Base length of 1-1/4 in
  • Base width of 1-1/4 in
  • Base height of 1-3/4 in
  • Post height of 5 in
  • Holders included
Catalog No. 50-005-29556