Learn More
MSC TESA Brown & Sharpe Outside Micrometer, 3-4 in, 0.0001 in Resolution, Vernier
Supplier: MSC 99870248
This mechanical outside micrometer is designed for precise external measurements. It offers a measuring range of 3 to 4 inches with a resolution of 0.0001 inch, utilizing a Vernier display for clear and accurate readings. This tool is built to industry standards and includes essential certifications for reliability.
- Measures external dimensions from 3 to 4 inches
- Provides a fine resolution of 0.0001 inch
- Ensures accuracy of +/-0.0001 inch
- Features a Vernier mechanical display
- Equipped with a fixed, friction thimble
- Has carbide tipped anvil and a flat spindle
- Comes with a fitted carrying case for protection
- Conforms to ISO 3611 and DIN 863-T3 standards
- Includes an inspection report and declaration of conformity