Learn More
Cayman Chemical TesaglItazar 10mg
SDP
Supplier:  Cayman Chemical 1679110 MG
Encompass_Preferred
A dual agonist of PPARα and γ (IC50s = 1 and 0.2 µM, respectively) that at 3 µM/kg/day for 3 weeks has been used to reduce insulin resistance in obese Zucker rats
Catalog No. 50-298-0395