Learn More
Medchemexpress LLC Tesirine intermediate-2 | 1430738-34-3 | 99.4% | 793.98 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY2414450MG
Encompass_Preferred
Tesirine intermediate-2 is an intermediate of Tesirine. Tesirine is a pyrrole benzodiazepine (PBD) dimer, which acts as a DNA small channel crosslinker with potent cytotoxicity. It is utilized in the synthesis of Antibody-Drug Conjugates (ADCs), where its warhead component is known for its strong anticancer cell activity. The product appears as a white to off-white solid. It is recommended for research use only and not intended for human use.
  • Intermediate of Tesirine
  • Utilized in the synthesis of antibody-drug conjugates (ADCs)
  • Acts as a DNA small channel crosslinker
  • Potent cytotoxicity
  • Warhead component is known for strong anticancer cell activity
  • Appears as a white to off-white solid
  • Recommended for research use only
Catalog No. 50-004-03462