Learn More
Grainger Test Cap Polysulfone
Supplier:  Grainger 16T766
Encompass_Preferred
The Oatey 16T766 Test Cap is a 1-1/2 inch PVC test cap designed for use on DWV piping systems. It features solvent weld sockets and is made of PVC plastic to provide chemical and corrosion resistance. This cap allows access for testing and inspection of piping systems up to 100 PSI. It can also be used for cleanouts. The dimensions are 2-1/2 inches length x 1-1/2 inches socket diameter. This product meets ASTM D2665 standards. Made in the USA.
Catalog No. 50-276-5412