Learn More
Cliawaived Inc 1 Panel Urine DipStick Test for Oxycodone
SDP
Supplier:  Cliawaived Inc CLIAIDTCII23
Encompass_Preferred
(50/Pack) CLIA waived single urine dip stick test for Oxycodone. Read results from 5-60 minutes. New hermetically sealed plastic housing prevents dip card from flooding. Easy to read. Negative results as soon as 1 minute. Offers improved accuracy and sensitivity testing.
Catalog No. NC1543101