Learn More
World Precision Instrument Test Electrode for EVOM2
SDP
Supplier:  World Precision Instrument 91750
Encompass
91750 is a 1000 ohm test resistor for use with the EVOM2 meter. Simply plug the electrode into the INPUT port and set the Function switch to OHMS. A properly calibrated meter will display 1000 ohms.
Catalog No. NC0539829