Learn More
Hach Company Ozone Test Kit, AccuVac Color Disc, HR 0.1-1.50 mg/L
Supplier:  Hach Company 2518050
Encompass
Fast, accurate visual method for determining ozone concentrations. Use to control disinfectant levels in water, wastewater, and industrial water. Kit contains 25 tests.
Catalog No. 50-199-6173