Learn More
Hach Company Chromium Test Kit, Model CH-14
Supplier:  Hach Company 222702
Encompass
Widely used as a corrosion inhibitor in cooling towers and engine cooling systems, chromium is also used in surface finishing, leather tanning, and other industries. The most widely used form, hexavalent chromium, is very toxic and strict limits are placed on discharges. Utilizing the drop count titration method to determine chromium (high range), this kit contains 100 tests.
Catalog No. 50-199-4731