Learn More
Hach Company Copper (Free & Total) Color Disc Test Kit, Model CU-6
Supplier:  Hach Company 2194100
Encompass
Copper may exist in natural waters and effluents as a soluble salt or as suspended solids. A small amount is essential for plants and animals, and concentrations exceeding 0.1 mg/L are useful for controlling algae and plankton growth. Quantities ranging from 0.02 to 0.1 mg/L are toxic for some fish, so its use for treating fish disease requires careful monitoring. Range: 0.1-4 mg/L; Contains 50 Free and 50 Total tests.
Catalog No. 50-199-5541