Learn More
Hach Company Hydrogen Sulfide Color Disc Test Kit, Model HS-WR
Supplier:  Hach Company 223801
Encompass
Resulting from the anaerobic decomposition of organic matter, hydrogen sulfide is present in many water supplies. Sewage and industrial wastes are other sources of sulfide pollution. Highly toxic, this compound has a characteristic rotten egg odor, which can be detected long before harmful concentrations are reached. Sulfide reacts with colorimetric reagents to form a blue color, which is measured with a color disc.
Catalog No. 50-199-4742