Learn More
Hach Company Phenols Test Kit, Model PL-1
Supplier:  Hach Company 2483600
Encompass
Utilizing the simple color disc method with a range of 0 - 4 mg/L, this kit contains 100 tests.
Catalog No. 50-199-6054