Learn More
Hach Company Phosphorus, Orthophosphate (Reactive) Test Kit, Model PO-19
Supplier:  Hach Company 224800
Encompass
Method / Chemistry: Color disc / Ascorbic acid. Ranges (mg/L): 0-0.8, 0-4, 0-40. Approx. Number of Tests: 100.
Catalog No. 50-199-4745