Learn More
Hach Company Surface Water Test Kit
Supplier:  Hach Company 2559800
Encompass
The ideal tool for source water monitoring and wastewater treatment plants, this kit uses drop count titration and color disc methods to provide economical, easy-to-use tests for ammonia (0-2.4 mg/L), chlorine (0-3.4 mg/L), pH (0-14), nitrate (0-40 mg/L), dissolved oxygen (0.2-20 mg/L), phosphorus (0-40 mg/L), and temperature. The rugged, lightweight kit includes the convenient pH Pocket Pro™ Tester and all necessary apparatus and reagents for testing.
Catalog No. 50-199-6230